Assay Details
| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172894.2 | NP_766482.2 | 20 - 21 | 2792 | 70 | |
| GenBank mRNA | AK075825.1 | - | 4 - 5 | 504 | 70 | |
| AK122450.1 | - | 16 - 17 | 3896 | 70 | ||
| BC053076.1 | - | 20 - 21 | 2792 | 5698212 | 70 | |
| BC141039.1 | - | 19 - 20 | 2339 | 9055651 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp6r1
Genomic Map