Assay Details
| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172894.2 | NP_766482.2 | 2 - 3 | 701 | 90 | |
| GenBank mRNA | AK080891.1 | - | 2 - 3 | 742 | 90 | |
| AK122450.1 | - | 2 - 3 | 701 | 90 | ||
| BC053076.1 | - | 2 - 3 | 701 | 5698212 | 90 | |
| BC141039.1 | - | 1 - 2 | 248 | 9055651 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp6r1
Genomic Map