Assay Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011958.3 | NP_036088.3 | 10 - 11 | 1009 | 121 | |
| GenBank mRNA | AJ003140.1 | - | 10 - 11 | 970 | 121 | |
| AK010258.1 | - | 10 - 11 | 993 | 121 | ||
| AK076324.1 | - | 10 - 11 | 1010 | 121 | ||
| AK169291.1 | - | 10 - 11 | 993 | 121 | ||
| BC015072.1 | - | 10 - 11 | 981 | 4212266 | 121 | |
| BC019748.1 | - | 10 - 11 | 951 | 5007346 | 121 | |
| Y16386.1 | - | 9 - 10 | 841 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Orc4
Genomic Map