Assay Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177313.1 | NP_001170784.1 | 3 - 4 | 291 | 121 | |
| NM_011958.3 | NP_036088.3 | 3 - 4 | 291 | 121 | ||
| NR_033442.1 | 3 - 4 | 291 | 121 | |||
| GenBank mRNA | AJ003140.1 | - | 3 - 4 | 252 | 121 | |
| AK010258.1 | - | 3 - 4 | 275 | 121 | ||
| AK011550.1 | - | 3 - 4 | 257 | 121 | ||
| AK012669.1 | - | 3 - 4 | 259 | 121 | ||
| AK049534.1 | - | 3 - 4 | 259 | 121 | ||
| AK076324.1 | - | 3 - 4 | 292 | 121 | ||
| AK084326.1 | - | 3 - 4 | 260 | 121 | ||
| AK145454.1 | - | 3 - 4 | 302 | 121 | ||
| AK169291.1 | - | 3 - 4 | 275 | 121 | ||
| BC015072.1 | - | 3 - 4 | 263 | 4212266 | 121 | |
| BC019748.1 | - | 3 - 4 | 233 | 5007346 | 121 | |
| Y16386.1 | - | 2 - 3 | 123 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Orc4
Genomic Map