Assay Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177313.1 | NP_001170784.1 | 4 - 5 | 379 | 118 | |
| NM_011958.3 | NP_036088.3 | 4 - 5 | 379 | 118 | ||
| NR_033442.1 | 4 - 5 | 379 | 118 | |||
| GenBank mRNA | AJ003140.1 | - | 4 - 5 | 340 | 118 | |
| AK010258.1 | - | 4 - 5 | 363 | 118 | ||
| AK011550.1 | - | 4 - 5 | 345 | 118 | ||
| AK012669.1 | - | 4 - 5 | 347 | 118 | ||
| AK049534.1 | - | 4 - 5 | 347 | 118 | ||
| AK076324.1 | - | 4 - 5 | 380 | 118 | ||
| AK084326.1 | - | 4 - 5 | 348 | 118 | ||
| AK145454.1 | - | 4 - 5 | 390 | 118 | ||
| AK169291.1 | - | 4 - 5 | 363 | 118 | ||
| BC015072.1 | - | 4 - 5 | 351 | 4212266 | 118 | |
| BC019748.1 | - | 4 - 5 | 321 | 5007346 | 118 | |
| Y16386.1 | - | 3 - 4 | 211 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Orc4
Genomic Map