Assay Details
| Gene Symbol: Sp3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045269.2 | 1 - 2 | 162 | 158 | ||
| GenBank mRNA | AK005870.1 | - | 1 - 2 | 162 | 158 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183265.1 | - | 162 | Chr. - - [-] on Build | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005870.1 transcripts.
Set Membership: |
|

Sp3os
Genomic Map