Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200011269.mm.1 |
NM_001289742.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011269.mm.1 |
NM_001289743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011269.mm.1 |
NM_178725.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011269.mm.1 |
TC0200001472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011265.mm.1 |
NM_001289744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011265.mm.1 |
NM_178725.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011265.mm.1 |
TC0200001472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011266.mm.1 |
NM_001289744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011266.mm.1 |
NM_178725.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011266.mm.1 |
TC0200001472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011267.mm.1 |
NM_001289744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011267.mm.1 |
NM_178725.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011267.mm.1 |
TC0200001472.mm.1 |
MTA TC |