Assay Details
| Gene Symbol: 8430437L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040503.1 | 4 - 5 | 832 | 93 | ||
| GenBank mRNA | AK018472.1 | - | 4 - 5 | 832 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle associated 7 | Cdca7 |
2310021G01Rik, AU044109, JPO1 |
-227.4kb | Sense | Chr. 2 - 72476189 - 72486892 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 6430710C18 gene | 6430710C18Rik | 5033406J01Rik | 21.2kb | Anti-sense | Chr. 2 - 72745846 - 72813717 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

8430437L04Rik
Genomic Map