Assay Details
| Gene Symbol: 4631405J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110333.1 | 4 - 5 | 575 | 74 | ||
| GenBank mRNA | AK028444.1 | - | 2287 | 74 | ||
| BC100298.1 | - | 4 - 5 | 575 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

4631405J19Rik
Genomic Map