Assay Details
| Gene Symbol: BC016548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC096650.1 | - | 1 - 2 | 48 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A930006I01 gene | A930006I01Rik | -15.3kb | Sense | Chr. 2 - 103338601 - 103372764 on NCBI Build GRCm38 | |
| Downstream | E74-like factor 5 | Elf5 |
ESE-2, ESE-5, ESE-5. |
22.6kb | Anti-sense | Chr. 2 - 103411693 - 103450989 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

BC016548
Genomic Map