Assay Details
| Gene Symbol: Phf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109690.1 | NP_001103160.1 | 14 - 15 | 1707 | 106 | |
| NM_001109691.1 | NP_001103161.1 | 17 - 18 | 1863 | 106 | ||
| NM_138755.2 | NP_620094.2 | 16 - 17 | 1725 | 106 | ||
| GenBank mRNA | AB105178.1 | - | 16 - 17 | 1631 | 106 | |
| AK037174.1 | - | 17 - 18 | 1864 | 106 | ||
| AK129424.1 | - | 16 - 17 | 3121 | 106 | ||
| AK137085.1 | - | 13 - 14 | 1674 | 106 | ||
| BC019181.1 | - | 16 - 17 | 1580 | 5008954 | 106 | |
| CX220811.1 | - | 4 - 5 | 413 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phf21a
Genomic Map