Assay Details
| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011957.2 | NP_036087.2 | 2 - 3 | 397 | 146 | |
| GenBank mRNA | AB017614.1 | - | 2 - 3 | 359 | 146 | |
| AK142368.1 | - | 398 | 146 | |||
| BC016447.1 | - | 2 - 3 | 372 | 4921584 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l1
Genomic Map