Assay Details
| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009963.4 | NP_034093.1 | 8 - 9 | 1388 | 96 | |
| GenBank mRNA | AB003433.1 | - | 8 - 9 | 1273 | 96 | |
| AF156987.1 | - | 8 - 9 | 1356 | 96 | ||
| AK041696.1 | - | 8 - 9 | 1356 | 96 | ||
| AK133781.1 | - | 8 - 9 | 1363 | 96 | ||
| AK172994.1 | - | 8 - 9 | 1320 | 96 | ||
| BC006077.1 | - | 2 - 3 | 296 | 96 | ||
| BC054794.1 | - | 8 - 9 | 1344 | 6834104 | 96 | |
| BC066799.1 | - | 8 - 9 | 1347 | 30103169 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cry2
Genomic Map