Assay Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080754.1 | NP_001074223.1 | 14 - 15 | 2910 | 127 | |
| NM_172669.3 | NP_766257.3 | 13 - 14 | 3183 | 127 | ||
| GenBank mRNA | AK129433.1 | - | 9 - 10 | 2447 | 127 | |
| AK148887.1 | - | 13 - 14 | 2815 | 127 | ||
| AK155513.1 | - | 13 - 14 | 3146 | 127 | ||
| AK169280.1 | - | 14 - 15 | 2863 | 127 | ||
| BC032293.1 | - | 14 - 15 | 2833 | 127 | ||
| BC054082.1 | - | 13 - 14 | 3116 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ambra1
Genomic Map