Assay Details
| Gene Symbol: Adssl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007421.2 | NP_031447.1 | 9 - 10 | 1017 | 79 | |
| GenBank mRNA | AK089520.1 | - | 9 - 10 | 992 | 79 | |
| AK146907.1 | - | 9 - 10 | 1020 | 79 | ||
| AK149634.1 | - | 9 - 10 | 992 | 79 | ||
| AK150874.1 | - | 9 - 10 | 988 | 79 | ||
| BC039943.1 | - | 9 - 10 | 1048 | 4014672 | 79 | |
| M74495.1 | - | 9 - 10 | 975 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adssl1
Genomic Map