| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200009137.mm.1 |
NM_001024602.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009137.mm.1 |
NM_001024602.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009137.mm.1 |
TC1200001269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200009140.mm.1 |
NM_001024602.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009140.mm.1 |
NM_001024602.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009140.mm.1 |
TC1200001269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200009139.mm.1 |
NM_001024602.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009139.mm.1 |
NM_001024602.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009139.mm.1 |
TC1200001269.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200009138.mm.1 |
NM_001024602.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009138.mm.1 |
NM_001024602.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200009138.mm.1 |
TC1200001269.mm.1 |
MTA TC |