Assay Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024602.3 | NP_001019773.2 | 3 - 4 | 424 | 70 | |
| GenBank mRNA | AK122236.1 | - | 4 - 5 | 353 | 70 | |
| AK147410.1 | - | 3 - 4 | 365 | 70 | ||
| BC050077.1 | - | 3 - 4 | 258 | 70 | ||
| BC141174.1 | - | 2 - 3 | 224 | 9055786 | 70 | |
| CJ125191.1 | - | 3 - 4 | 425 | 70 | ||
More Information
Set Membership: |
|

Cep170b
Genomic Map