Assay Details
| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144884.2 | NP_659133.1 | 2 - 3 | 513 | 64 | |
| GenBank mRNA | AJ298841.1 | - | 2 - 3 | 468 | 64 | |
| AK038891.1 | - | 2 - 3 | 471 | 64 | ||
| AK079728.1 | - | 1 - 2 | 785 | 64 | ||
| AK082936.1 | - | 2 - 3 | 423 | 64 | ||
| AK160361.1 | - | 2 - 3 | 469 | 64 | ||
| BC017683.1 | - | 2 - 3 | 493 | 4238480 | 64 | |
| BY724297.1 | - | 2 - 3 | 502 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tor1a
Genomic Map