| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200017399.mm.1 |
NM_030135.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017399.mm.1 |
NM_030135.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017399.mm.1 |
TC0200002199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200017398.mm.1 |
NM_030135.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017398.mm.1 |
NM_030135.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017398.mm.1 |
TC0200002199.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200017400.mm.1 |
NM_030135.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017400.mm.1 |
NM_030135.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200017400.mm.1 |
TC0200002199.mm.1 |
MTA TC |