Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300008952.mm.1 |
NM_001199141.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008952.mm.1 |
NM_144516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008952.mm.1 |
TC1300001424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008950.mm.1 |
NM_001199141.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008950.mm.1 |
NM_144516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008950.mm.1 |
TC1300001424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008953.mm.1 |
NM_001199141.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008953.mm.1 |
NM_144516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008953.mm.1 |
TC1300001424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008954.mm.1 |
NM_001199141.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008954.mm.1 |
NM_144516.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008954.mm.1 |
TC1300001424.mm.1 |
MTA TC |