| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300009234.mm.1 |
NM_001199043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009234.mm.1 |
NM_001199043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009234.mm.1 |
NM_018886.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009234.mm.1 |
NM_018886.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009234.mm.1 |
TC1300001448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009229.mm.1 |
NM_001199043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009229.mm.1 |
NM_001199043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009229.mm.1 |
NM_018886.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009229.mm.1 |
NM_018886.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009229.mm.1 |
TC1300001448.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009230.mm.1 |
NM_001199043.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009230.mm.1 |
NM_001199043.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009230.mm.1 |
NM_018886.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009230.mm.1 |
NM_018886.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009230.mm.1 |
TC1300001448.mm.1 |
MTA TC |