Assay Details
| Gene Symbol: Elac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023479.2 | NP_075968.2 | 9 - 10 | 854 | 82 | |
| GenBank mRNA | AF308696.2 | - | 9 - 10 | 792 | 82 | |
| AK156730.1 | - | 9 - 10 | 855 | 82 | ||
| AK172604.1 | - | 8 - 9 | 1223 | 82 | ||
| BC023134.1 | - | 9 - 10 | 798 | 82 | ||
| BC048235.1 | - | 9 - 10 | 788 | 6493124 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elac2
Genomic Map