Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028316.3 | NP_082592.2 | 1 - 2 | 59 | 74 | |
| GenBank mRNA | AK169973.1 | - | 1 - 2 | 71 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK169973.1, NM_028316.3 transcripts.
Set Membership: |
|

Zfp444
Genomic Map