Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146024.1 | NP_001139496.1 | 3 - 4 | 524 | 124 | |
| NM_028316.3 | NP_082592.2 | 2 - 3 | 404 | 124 | ||
| GenBank mRNA | AK018079.1 | - | 3 - 4 | 526 | 124 | |
| AK089886.1 | - | 2 - 3 | 373 | 124 | ||
| AK169973.1 | - | 2 - 3 | 416 | 124 | ||
| BC023396.2 | - | 3 - 4 | 590 | 124 | ||
| BC037180.1 | - | 2 - 3 | 372 | 124 | ||
| BC139359.1 | - | 2 - 3 | 360 | 8862381 | 124 | |
| BC139361.1 | - | 2 - 3 | 360 | 8862383 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp444
Genomic Map