Assay Details
| Gene Symbol: Dclre1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110214.1 | NP_001103684.1 | 3 - 4 | 329 | 103 | |
| NM_146114.3 | NP_666226.2 | 3 - 4 | 329 | 103 | ||
| NM_175683.4 | NP_783614.1 | 3 - 4 | 329 | 103 | ||
| GenBank mRNA | AF387731.1 | - | 3 - 4 | 311 | 103 | |
| AK037126.1 | - | 3 - 4 | 296 | 103 | ||
| AK038788.1 | - | 3 - 4 | 259 | 103 | ||
| AK052369.1 | - | 3 - 4 | 285 | 103 | ||
| AK052490.1 | - | 3 - 4 | 268 | 103 | ||
| AK166324.1 | - | 3 - 4 | 272 | 103 | ||
| BC108935.2 | - | 3 - 4 | 282 | 40039591 | 103 | |
| BY729142.1 | - | 3 - 4 | 312 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dclre1c
Genomic Map