Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_001110214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_001110214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_146114.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_146114.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_175683.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
NM_175683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000069.mm.1 |
TC0200000006.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_001110214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_001110214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_146114.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_146114.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_175683.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
NM_175683.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000067.mm.1 |
TC0200000006.mm.1 |
MTA TC |