Assay Details
| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163394.1 | NP_001156866.1 | 11 - 12 | 1574 | 83 | |
| NM_001163396.2 | NP_001156868.1 | 12 - 13 | 1348 | 83 | ||
| GenBank mRNA | AF279662.1 | - | 10 - 11 | 1085 | 83 | |
| AK137823.1 | - | 11 - 12 | 1164 | 83 | ||
| AK151633.1 | - | 13 - 14 | 1350 | 83 | ||
| BC096017.1 | - | 11 - 12 | 1467 | 83 | ||
More Information
Set Membership: |
|

Evl
Genomic Map