Assay Details
| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164488.1 | NP_001157960.1 | 1 - 2 | 39 | 61 | |
| NM_011710.3 | NP_035840.3 | 1 - 2 | 52 | 61 | ||
| GenBank mRNA | AK004541.1 | - | 1 - 2 | 40 | 61 | |
| BC003450.1 | - | 1 - 2 | 38 | 2654231 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004541.1, BC003450.1, NM_001164488.1, NM_011710.3 transcripts.
Set Membership: |
|

Wars
Genomic Map