Assay Details
| Gene Symbol: Vrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029843.1 | NP_001025014.1 | 12 - 13 | 1257 | 73 | |
| GenBank mRNA | AF080252.1 | - | 11 - 12 | 1170 | 73 | |
| AK032344.1 | - | 12 - 13 | 1228 | 73 | ||
| AK088230.1 | - | 12 - 13 | 1181 | 73 | ||
| AK166676.1 | - | 12 - 13 | 1225 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vrk1
Genomic Map