Assay Details
| Gene Symbol: Clmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053155.2 | NP_444385.2 | 12 - 13 | 3021 | 91 | |
| NR_104435.1 | 13 - 14 | 3102 | 91 | |||
| NR_104438.1 | 13 - 14 | 3157 | 91 | |||
| GenBank mRNA | AB059644.1 | - | 12 - 13 | 3014 | 91 | |
| AB059647.1 | - | 13 - 14 | 3095 | 91 | ||
| AB059648.1 | - | 13 - 14 | 3150 | 91 | ||
| AK077023.1 | - | 9 - 10 | 2603 | 91 | ||
| BC141280.1 | - | 12 - 13 | 2954 | 9055892 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clmn
Genomic Map