Assay Details
| Gene Symbol: Nqo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020282.3 | NP_064678.1 | 2 - 3 | 229 | 134 | |
| GenBank mRNA | BC027629.1 | - | 2 - 3 | 137 | 1225122 | 134 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027629.1, NM_020282.3 transcripts.
Set Membership: |
|

Nqo2
Genomic Map