Assay Details
| Gene Symbol: Nqo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163239.1 | NP_001156711.1 | 2 - 3 | 310 | 87 | |
| NM_001163241.1 | NP_001156713.1 | 3 - 4 | 398 | 87 | ||
| NM_001163242.1 | NP_001156714.1 | 3 - 4 | 327 | 87 | ||
| NM_020282.3 | NP_064678.1 | 4 - 5 | 480 | 87 | ||
| GenBank mRNA | AF252260.1 | - | 3 - 4 | 326 | 87 | |
| AK008138.1 | - | 2 - 3 | 282 | 87 | ||
| AK008465.1 | - | 2 - 3 | 285 | 87 | ||
| AK010842.1 | - | 3 - 4 | 370 | 87 | ||
| AK049781.1 | - | 3 - 4 | 336 | 87 | ||
| BC027629.1 | - | 4 - 5 | 388 | 1225122 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nqo2
Genomic Map