Assay Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007772.2 | NP_031798.2 | 1 - 2 | 63 | 98 | |
| GenBank mRNA | AK039808.1 | - | 1 - 2 | 63 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039808.1, NM_007772.2 transcripts.
Set Membership: |
|

Hivep1
Genomic Map