Assay Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007772.2 | NP_031798.2 | 4 - 5 | 6204 | 114 | |
| GenBank mRNA | AK134323.1 | - | 1 - 2 | 596 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hivep1
Genomic Map