Assay Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007772.2 | NP_031798.2 | 7 - 8 | 6615 | 61 | |
| GenBank mRNA | AK134323.1 | - | 4 - 5 | 1007 | 61 | |
| AK136419.1 | - | 381 | 61 | |||
| X68946.1 | - | 3 - 4 | 375 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hivep1
Genomic Map