Assay Details
| Gene Symbol: Cap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026056.4 | NP_080332.1 | 6 - 7 | 838 | 67 | |
| GenBank mRNA | AK010235.1 | - | 6 - 7 | 585 | 67 | |
| AK013331.1 | - | 6 - 7 | 595 | 67 | ||
| AK014979.1 | - | 2 - 3 | 234 | 67 | ||
| AK033071.1 | - | 6 - 7 | 594 | 67 | ||
| AK047701.1 | - | 4 - 5 | 405 | 67 | ||
| AK076737.1 | - | 2 - 3 | 258 | 67 | ||
| BC050752.1 | - | 6 - 7 | 599 | 6533812 | 67 | |
| BC057937.1 | - | 6 - 7 | 653 | 5325599 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cap2
Genomic Map