Assay Details
| Gene Symbol: Cap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026056.4 | NP_080332.1 | 9 - 10 | 1309 | 99 | |
| GenBank mRNA | AK010235.1 | - | 9 - 10 | 1056 | 99 | |
| AK013331.1 | - | 9 - 10 | 1066 | 99 | ||
| AK033071.1 | - | 9 - 10 | 1065 | 99 | ||
| AK035556.1 | - | 9 - 10 | 1310 | 99 | ||
| AK047701.1 | - | 7 - 8 | 876 | 99 | ||
| AK049813.1 | - | 6 - 7 | 752 | 99 | ||
| AK076737.1 | - | 5 - 6 | 729 | 99 | ||
| BC050752.1 | - | 9 - 10 | 1070 | 6533812 | 99 | |
| BC057937.1 | - | 9 - 10 | 1124 | 5325599 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cap2
Genomic Map