Assay Details
| Gene Symbol: Cap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026056.4 | NP_080332.1 | 10 - 11 | 1433 | 112 | |
| GenBank mRNA | AK010235.1 | - | 10 - 11 | 1180 | 112 | |
| AK013331.1 | - | 10 - 11 | 1190 | 112 | ||
| AK014979.1 | - | 6 - 7 | 828 | 112 | ||
| AK033071.1 | - | 10 - 11 | 1189 | 112 | ||
| AK035556.1 | - | 10 - 11 | 1434 | 112 | ||
| AK047701.1 | - | 8 - 9 | 1000 | 112 | ||
| AK049813.1 | - | 7 - 8 | 876 | 112 | ||
| AK076737.1 | - | 6 - 7 | 853 | 112 | ||
| BC050752.1 | - | 10 - 11 | 1194 | 6533812 | 112 | |
| BC057937.1 | - | 10 - 11 | 1248 | 5325599 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cap2
Genomic Map