Assay Details
| Gene Symbol: Gm4872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073371.1 | 2 - 3 | 206 | 84 | ||
| GenBank mRNA | AK052410.1 | - | 2 - 3 | 207 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | JAZF zinc finger 1 | Jazf1 |
AI591476, C820002C15, Jaz1, Tip27 |
-153.3kb | Anti-sense | Chr. 6 - 52768065 - 53068624 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9430076C15 gene | 9430076C15Rik | 62.3kb | Sense | Chr. 6 - 53287295 - 53397216 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm4872
Genomic Map