| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600005110.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005110.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005110.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005106.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005106.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005106.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005108.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005108.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005108.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005107.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005107.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005107.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005109.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005109.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005109.mm.1 |
NM_001142781.1 |
Refseq |