Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142781.1 | NP_001136253.1 | 16 - 17 | 2241 | 80 | |
| GenBank mRNA | AK142385.1 | - | 16 - 17 | 2159 | 80 | |
| AK161304.1 | - | 16 - 17 | 81 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Fam188b
Genomic Map