Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142781.1 | NP_001136253.1 | 17 - 18 | 2310 | 65 | |
| GenBank mRNA | AK142385.1 | - | 17 - 18 | 2228 | 65 | |
| AK161304.1 | - | 17 - 18 | 2190 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam188b
Genomic Map