Assay Details
| Gene Symbol: 5430402O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015581.1 | 2 - 3 | 442 | 137 | ||
| GenBank mRNA | AK017262.1 | - | 2 - 3 | 137 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6371 | Mir6371 |
Gm27376, mmu-mir-6371 |
-3.0kb | Sense | Chr. 6 - 50572398 - 50572508 on NCBI Build GRCm38 |
| Downstream | neuropeptide VF precursor | Npvf | Rfrp | 60.9kb | Anti-sense | Chr. 6 - 50650871 - 50654393 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5430402O13Rik
Genomic Map