Assay Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198977.1 | NP_001185906.1 | 9 - 10 | 1378 | 88 | |
| NM_011518.2 | NP_035648.2 | 9 - 10 | 1382 | 88 | ||
| GenBank mRNA | AK049207.1 | - | 9 - 10 | 1407 | 88 | |
| AK136363.1 | - | 9 - 10 | 1381 | 88 | ||
| AK143564.1 | - | 9 - 10 | 1411 | 88 | ||
| BC065121.1 | - | 9 - 10 | 1314 | 30533300 | 88 | |
| U25685.1 | - | 8 - 9 | 1166 | 88 | ||
| U36776.1 | - | 9 - 10 | 1385 | 88 | ||
| Z49877.1 | - | 9 - 10 | 1640 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syk
Genomic Map