Assay Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198977.1 | NP_001185906.1 | 7 - 8 | 1112 | 87 | |
| NM_011518.2 | NP_035648.2 | 7 - 8 | 1116 | 87 | ||
| GenBank mRNA | AK136363.1 | - | 7 - 8 | 1115 | 87 | |
| AK143564.1 | - | 7 - 8 | 1145 | 87 | ||
| BC065121.1 | - | 7 - 8 | 1048 | 30533300 | 87 | |
| U25685.1 | - | 6 - 7 | 900 | 87 | ||
| U36776.1 | - | 7 - 8 | 1119 | 87 | ||
| Z49877.1 | - | 7 - 8 | 1377 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syk
Genomic Map