Assay Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301341.1 | NP_001288270.1 | 1 - 2 | 76 | 61 | |
| GenBank mRNA | AK078777.1 | - | 1 - 2 | 77 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078777.1, NM_001301341.1 transcripts.
Set Membership: |
|

Hnrnpk
Genomic Map