Assay Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301341.1 | NP_001288270.1 | 2 - 3 | 196 | 168 | |
| GenBank mRNA | AK078777.1 | - | 2 - 3 | 197 | 168 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078777.1, NM_001301341.1 transcripts.
Set Membership: |
|

Hnrnpk
Genomic Map