Assay Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022724.4 | NP_073561.2 | 3 - 4 | 1092 | 131 | |
| NR_027509.1 | 2 - 3 | 946 | 131 | |||
| GenBank mRNA | AK015728.1 | - | 1 - 2 | 138 | 131 | |
| AK045206.1 | - | 2 - 3 | 930 | 131 | ||
| AK083457.1 | - | 3 - 4 | 1070 | 131 | ||
| AK165278.1 | - | 2 - 3 | 745 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Suv39h2
Genomic Map