Assay Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022724.4 | NP_073561.2 | 4 - 5 | 1243 | 99 | |
| NR_027509.1 | 3 - 4 | 1097 | 99 | |||
| GenBank mRNA | AF149205.1 | - | 4 - 5 | 1221 | 99 | |
| AK015728.1 | - | 2 - 3 | 289 | 99 | ||
| AK045206.1 | - | 3 - 4 | 1081 | 99 | ||
| AK083457.1 | - | 4 - 5 | 1221 | 99 | ||
| AK165278.1 | - | 3 - 4 | 896 | 99 | ||
| BC032960.1 | - | 3 - 4 | 566 | 1330053 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Suv39h2
Genomic Map