Assay Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015765.2 | NP_056580.2 | 8 - 9 | 855 | 71 | |
| GenBank mRNA | AK031489.1 | - | 794 | 71 | ||
| AK145937.1 | - | 8 - 9 | 809 | 71 | ||
| AK153397.1 | - | 8 - 9 | 817 | 71 | ||
| BU505607.1 | - | 5 - 6 | 506 | 71 | ||
| U08215.1 | - | 8 - 9 | 815 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa14
Genomic Map